N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: DABCYL acid, SE, DABCYL N-Succinimidyl Ester, 4-[4-(Dimethylamino)phenylazo]benzoic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 4-((4-(dimethylamino)phenyl)diazenyl)benzoate

2004 2005
76481-304EA 437.45 USD
76481-304 76481-306
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥95%
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate

DABCYL, SE is the amino-reactive form of DABCYL, and widely used to prepare a variety of FRET-based probes that contain DABCYL.

Formula: C₁₉H₁₈N₄O₄
MDL Number: MFCD00467415
CAS Number: 146998-31-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3288045e91764d21e55c94400a22af9c4d9c43f3&vid=18c9f4e9f08082b5d78265a3870e5b3b582b111d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR