7-Methoxycoumarin-3-carboxylic acid ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 7-Methoxy-2-oxo-2H-chromene-3-carboxylic acid

560 561
76480-984EA 188.87 USD
76480-984 76480-986
7-Methoxycoumarin-3-carboxylic acid ≥95%
7-Methoxycoumarin-3-carboxylic acid

7-Methoxycoumarin-3-carboxylic acid is used for HPLC derivatization.

Formula: C₁₁H₈O₅
MW: 220.18 g/mol
MDL Number: MFCD00452770
CAS Number: 20300-59-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=822555a76585018f1d64aa1962af306a89fbd630&vid=cd52cba0d62aab0731a70b771ec2c8f1fb6b2757&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR