7-Hydroxycoumarin-3-carboxylic acid ≥90%
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
Umbelliferone-3-carboxylic Acid, 7-Hydroxy-2-oxo-2H-chromene-3-carboxylic acid
549
550
76480-966EA
124.31
USD
76480-966
76480-968
7-Hydroxycoumarin-3-carboxylic acid ≥90%
7-Hydroxycoumarin-3-carboxylic acid
7-Hydroxycoumarin-3-carboxylic acid is a blue fluorophores for labeling proteins and nucleic acids mostly through the in situ formation of its succinimidyl ester catalyzed by EDAC.
Formula:
C₁₀H₆O₅ |
MDL Number:
MFCD00017491 CAS Number: 779-27-1 ADR: 5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...