5-Carboxy-X-rhodamine, NHS ester ≥90%
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
5-ROX, SE, 5-ROX N-succinimidyl ester
389
391
76480-864EA
249.58
USD
76480-864
76480-866
5-Carboxy-X-rhodamine, NHS ester ≥90%
5-Carboxy-X-rhodamine, NHS ester
Single isomers of ROX are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purifications that are often required in bioconjugations.
Formula:
C₃₇H₃₃N₃O₇ MW: 631.685 g/mol |
CAS Number:
209734-74-7 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...