6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) ≥90%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 6-Carboxytetramethylrhodamine N-succinimidyl ester, 6-TAMRA, SE, 6-TAMRA N-succinimidyl ester

376 377 378
76480-848EA 129.69 USD
76480-848 76480-850 76480-852
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) ≥90%
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE)

The single TAMRA isomers are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purification that is often required in the conjugation processes.

Formula: C₂₉H₂₅N₃O₇
CAS Number: 150810-69-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58266059a983bf709d98cefa90c90f15d1a35310&vid=87fef656765193135897bbee96ef4d73f827ba19&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR