6-TAMRA ≥95%

Supplier: AAT BIOQUEST INC
Warning

366 367 368
76480-830EA 96.26 USD
76480-830 76480-832 76480-834
6-TAMRA ≥95%
6-TAMRA

6-TAMRA is the purified single isomer of 5(6)-TAMRA.

Formula: C₂₅H₂₂N₂O₅
MDL Number: MFCD01073638
CAS Number: 91809-67-5
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=011fcf33fa44c268cdb7ddb21ea981c45e9f8fac&vid=0d2f9461e84df93296b0ae59e0bf2ca012454754&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR