5-Carboxyfluorescein ≥95%

Supplier: AAT BIOQUEST INC
Warning

103 104 105
76480-548EA 96.26 USD
76480-548 76480-550 76480-552
5-Carboxyfluorescein ≥95%
5-Carboxyfluorescein

5-FAM is the purified single isomer of carboxyfluorescein.

Formula: C₂₁H₁₂O₇.xH₂O
MDL Number: MFCD00036874
CAS Number: 76823-03-5
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cc80d5d97dac535b6103784dd6eed3b35f78805&vid=2db347ed8b66317c345cd473e54fe38375ec62f4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR