Indocyanine green ≥95%

Supplier: AAT BIOQUEST INC
Warning

91 96
76480-534EA 375.27 USD
76480-534 76480-536
Indocyanine green ≥95%
Indocyanine green

Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.

Formula: C₄₃H₄₇N₂NaO₆S₂
MDL Number: MFCD00013078
CAS Number: 3599-32-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e512f181aaa9b4841ceb4424b30925b04e6e09f8&vid=ec59cd880ff67badb50a7e9b0b8a7fb649f03517&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR