7-Amino-4-(trifluoromethyl)coumarin ≥95%

Supplier: AAT BIOQUEST INC
Warning

56 57 58
76480-492EA 124.31 USD
76480-492 76480-494 76480-496
7-Amino-4-(trifluoromethyl)coumarin ≥95%
7-Amino-4-(trifluoromethyl)coumarin

This compound is the precursor for a variety of AFC enzyme substrates.

Formula: C₁₀H₆F₃NO₂
MW: 229.16 g/mol
MDL Number: MFCD00006858
CAS Number: 53518-15-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=680d0dc32cd968a67451b82cb2ee8163e964cd3a&vid=bffdd07a825e9bbe13a96da150b6c5aac4c63dc5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR