1,3,4,6-Tetrachloro-3-6-diphenylglycouril

Synonyms: Iodo-Gen®, NSC 4462, 1,3,4,6-Tetrachloro-3α,6α-diphenylglycouril

M-Clarity™ quality level = MQ100

T0656-1G T0656-250MG
220225-696EA 592.12 USD
220225-696 220225-697
1,3,4,6-Tetrachloro-3-6-diphenylglycouril
1,3,4,6-Tetrachloro-3-6-diphenylglycouril

Iodination reagent used in radioiodination acting as an oxidative agent


Reagent involved in comparison studies with chloramine-T
Reagent used for oxidation of urazoles


Precaución: Iodo-Gen is a registered trademark of Pierce Biotechnology, Inc.

Formula: C₁₆H₁₀Cl₄N₄O₂
MW: 432.088 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00027367
CAS Number: 51592-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR