Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) ≥98% (by HPLC), Sigma-Aldrich®

Synonyms: N-Hydroxy sulphosuccinimide sodium salt, Hydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium salt, N-Hydroxysulphosuccinimide sodium salt, Sulfo NHS, Sulpho-N-Hydroxysuccinimide

56485-250MG 56485-5G 56485-1G
220077-401EA 126 USD
220077-401 220077-402 220077-400
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) ≥98% (by HPLC), Sigma-Aldrich®
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt)

N-Hydroxysulfosuccinimide sodium salt is a water-soluble sulfonated analog of N-hydroxysuccinimide. It can undergo a coupling reaction with carboxylic acids in the presence of carbodiimides such as 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide(EDC) to form hydrophilic N-hydroxysulfosuccinimide active esters. These esters are useful in protein cross-linking experiments.

Formula: C₄H₄NNaO₆S
MW: 217.13 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00043100
CAS Number: 106627-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR