HyClone™ Newborn Bovine Calf Serum, New Zealand, HyClone products (Cytiva)
Supplier: Cytiva
3040101IR25-40 3040102IR25-40 3040101IR30-45 3040102IR30-45
76324-902EA 106.72 USD
76324-902 76324-904 76324-934 76324-936
HyClone™ Newborn Bovine Calf Serum, New Zealand, HyClone products (Cytiva)
Sera (Serum)
All New Zealand calf serum products are carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases. We follow the same standards in processing of our calf serum as with our Fetal Bovine Serum (FBS) to enable offering the highest quality and traceability.
  • A cost-effective alternative to Fetal Bovine Serum (FBS)
  • Sourced from New Zealand with complete traceability back to original source
  • Age of animal at time of collection less than 10 days old
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
New Zealand Newborn Bovine Calf Serum is filtered through sequential 100 nm (0.1 µm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.