bisBenzimide H 33342 trihydrochloride ≥95%, Ultra Pure Grade

Supplier: AAT BIOQUEST INC

17530 17533
76482-874EA 124.31 USD
76482-874 76482-876
bisBenzimide H 33342 trihydrochloride ≥95%, Ultra Pure Grade
bisBenzimide H 33342 trihydrochloride

The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy.


  • Bright Fluorescence
  • 95% Purity
  • Useful For Labeling Nuclei/Mitochondria
  • DAPI Alternative

Formula: C₂₇H₂₈N₆O·3HCl·xH₂O
MDL Number: MFCD00012678
CAS Number: 875756-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR