Sulforhodamine B-VAD-FMK (SRB-VAD-FMK) ≥90%
Supplier: AAT BIOQUEST INC
13472 13482
76482-152EA 124.31 USD
76482-152 76482-154
Sulforhodamine B-VAD-FMK (SRB-VAD-FMK) ≥90%
Sulforhodamine B-VAD-FMK (SRB-VAD-FMK)
SRB-VAD is a red fluorescent cell-permeable polycaspase inhibitor to target caspases 1, 2, 3, 6, 8, 9, or 10.
  • Caspase Inhibitor
  • High Intracellular Retention
  • Useful In Multiple Cell Lines/Species
Precaución: For Research Use Only
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26e08a12c235d02821c2026a59c3608596b3c70f&vid=7cb39c3565197d5a55e0af65d3784d493c8c5829&prid=web&clid=SAPDAM