FAM-VAD-FMK ≥95%
Supplier: AAT BIOQUEST INC
13470 13484
76482-146EA 124.31 USD
76482-146 76482-148
FAM-VAD-FMK ≥95%
FAM-VAD-FMK
FAM-VAD is a green fluorescent cell-permeable polycaspase inhibitor to target caspases 1, 2, 3, 6, 8, 9, or 10.
  • Caspase Inhibitor
  • High Intracellular Retention
  • Useful In Multiple Cell Lines/Species
Precaución: For Research Use Only
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ed03cd065307056b7d44ffb552b21e928a07986&vid=4eab14ed8b03ce805a4d09cbfbadbb557ecc5f7c&prid=web&clid=SAPDAM