iFluor® 568, Succinimidyl Ester
Supplier: AAT BIOQUEST INC
1049 71049 71522 71572
76484-378EA 249.58 USD
76484-378 76484-380 76484-382 76484-384
iFluor® 568, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Precaución: For Research Use Only