iFluor® 710, Succinimidyl Ester
Supplier: AAT BIOQUEST INC
1045 71045 71520 71570
76481-162EA 249.58 USD
76481-162 76481-164 76481-166 76481-168
iFluor® 710, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Precaución: For Research Use Only