iFluor® 750, Succinimidyl Ester
Supplier: AAT BIOQUEST INC
1037 71037 71515 71565
76481-138EA 249.58 USD
76481-138 76481-140 76481-142 76481-144
iFluor® 750, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are developed for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Precaución: For Research Use Only