iFluor® 700, Succinimidyl Ester
Supplier: AAT BIOQUEST INC
1036 71036 71514 71564
76481-130EA 249.58 USD
76481-130 76481-132 76481-134 76481-136
iFluor® 700, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Precaución: For Research Use Only