iFluor® 660, Succinimidyl Ester
Supplier: AAT BIOQUEST INC
1032 71032 71511 71561
76481-114EA 249.58 USD
76481-114 76481-116 76481-118 76481-120
iFluor® 660, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Precaución: For Research Use Only