iFluor® 555, Succinimidyl Ester
Supplier: AAT BIOQUEST INC
1028 71028 71507 71557
76481-082EA 249.58 USD
76481-082 76481-084 76481-086 76481-088
iFluor® 555, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Precaución: For Research Use Only