iFluor® 488, Succinimidyl Ester
Supplier: AAT BIOQUEST INC
1023 71023 71503 71553
76481-058EA 249.58 USD
76481-058 76481-060 76481-062 76481-064
iFluor® 488, Succinimidyl Ester
iFluor®
Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Precaución: For Research Use Only