iFluor® 405, Succinimidyl Ester
Supplier: AAT Bioquest
1021 71021 71501 71551
76481-050EA 249.58 USD
76481-050 76481-052 76481-054 76481-056
iFluor® 405, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Precaución: For Research Use Only