iFluor® 350, Succinimidyl Ester fluorescent probe
Supplier: AAT Bioquest
1020 71020 71500 71550
76481-042EA 249.58 USD
76481-042 76481-044 76481-046 76481-048
iFluor® 350, Succinimidyl Ester fluorescent probe
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Precaución: For Research Use Only