6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE) single isomer ≥90%

Supplier: AAT BIOQUEST INC

Synonyms: Carboxyrhodamine 6G succinimidyl ester, 6-isomer, 6-CR6G, NHS ester, 6-Carboxyrhodamine 6G, succinimidyl ester

342 346
76480-796EA 96.26 USD
76480-796 76480-798
6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE) single isomer ≥90%
6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE)

6-CR6G, SE is amine-reactive derivative of 6-CR6G.


  • Bright Fluorescence
  • High Photostability
  • Useful For Small Molecule Labeling
  • 90% Purity


Precaución: For Research Use Only

Formula: C₃₁H₂₉N₃O₇
CAS Number: 216699-37-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d36883ed84fd30ac7c988f5be9e3a418d8e6169b&vid=4b1164afe8415ba874706fbf10745e08f8686b8c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR