6-TAMRA alkyne ≥90%

Supplier: AAT Bioquest

491 966
76480-942EA 188.87 USD
76480-942 76480-944
6-TAMRA alkyne ≥90%
6-TAMRA alkyne

6-TAMRA alkyne is an excellent azide-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.


  • Bright Fluorescence
  • Click Chemistry Conjugation Reagent


Precaución: For Research Use Only

Formula: C28H25N3O4
CAS Number: 1352649-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bad9af9fa25bb0ac220268057f60f6d68983b995&vid=35d7061a4629f2d98630b464ab2c3a35b53fffe5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR