5-TAMRA alkyne ≥95%
Supplier: AAT BIOQUEST INC
487 961
76480-934EA 188.87 USD
76480-934 76480-936
5-TAMRA alkyne ≥95%
5-TAMRA alkyne
5-TAMRA alkyne is an excellent azide-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.
  • Bright Fluorescence
  • Click Chemistry Conjugation Reagent
Precaución: For Research Use Only
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28b4192865079abcc85874a09f3586600d1cec1f&vid=7444217d08247ac8bc7ec72a5447bdee3e3752d6&prid=web&clid=SAPDAM