5-TAMRA azide ≥95%
Supplier: AAT BIOQUEST INC
486 960
76480-930EA 188.87 USD
76480-930 76480-932
5-TAMRA azide ≥95%
5-TAMRA azide
5-TAMRA azide is an excellent alkyne-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.
  • Bright Fluorescence
  • Click Chemistry Conjugation Reagent
Precaución: For Research Use Only
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9be4581c739999595f0e9f6c3fb4e409a246315e&vid=351e6977658ab50d5ee61f684cc62d03db9d5e8f&prid=web&clid=SAPDAM