California Red™ SE ≥90% Superior Replacement to Texas Red®
Supplier: AAT BIOQUEST INC
473
479
76480-912EA
188.87
USD
76480-912
76480-914
California Red™ SE ≥90% Superior Replacement to Texas Red®
California Red™ SE
Although Texas Red® is the most popular labeling reagent of sulfonyl chloride, it is quite unstable in water.
- Identical spectra to Texas Red
- High Stability
- Improved Labeling Efficiency
Precaución: For Research Use Only