Sulfo-Cyanine 5 carboxylic acid ≥95%

Supplier: AAT BIOQUEST INC

266 267
76484-618EA 626.51 USD
76484-618 76484-620
Sulfo-Cyanine 5 carboxylic acid ≥95%
Sulfo-Cyanine 5 carboxylic acid

A variety of cyanine dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.


  • pH Tolerance From 3-10
  • Bright Fluorescence
  • Multiple Bioconjugation Applications


Precaución: For Research Use Only

Formula: C32H37N2NaO8S2
MW: 664.19 g/mol
CAS Number: 1121756-16-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8731290047d01203706cc0b747434c1801ae7206&vid=c4ac2e388b9713e51a8f44cec7fdc274ba92c526&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR