Sulfo-Cyanine 3 carboxylic acid

Supplier: AAT BIOQUEST INC

Synonyms: 2-[3-[1-(5-Carboxypentyl)-3,3-dimethyl-5-sulphoindolin-2-ylidene]prop-1-en-1-yl]-1-ethyl-3,3-dimethyl-3H-indol-1-ium-5-sulphonate, Cy3, 2-(3-(1-(5-Carboxypentyl)-3,3-dimethyl-5-sulphoindolin-2-ylidene)prop-1-en-1-yl)-1-ethyl-3,3-dimethyl-3H-indol-1-ium-5-sulphonate

260 261
76484-610EA 626.51 USD
76484-610 76484-612
Sulfo-Cyanine 3 carboxylic acid
Sulfo-Cyanine 3 carboxylic acid

A variety of cyanine dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.


  • Multiple Bioconjugation Applications
  • Excellent Quality


Precaución: For Research Use Only

Formula: C₃₁H₃₈N₂O₈S₂
MW: 630.77 g/mol
MDL Number: MFCD28978247
CAS Number: 146368-13-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caba19f351d1b7c2bfc876abc03ad12d0461c1f6&vid=1b8f296a7b6f51559f9237c40b6f3492273e6153&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR