FITC-xtra ≥90%
Supplier: AAT Bioquest
135 136
76484-418EA 249.58 USD
76484-418 76484-420
FITC-xtra ≥90%
FITC-xtra
Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.
  • pH-Independent Fluorescence
  • High Signal/Background Ratio
Precaución: For Research Use Only