6-FAM Alkyne ≥95%
Supplier: AAT BIOQUEST INC
134
956
76480-612EA
188.87
USD
76480-612
76480-614
6-FAM Alkyne ≥95%
6-FAM Alkyne
6-FAM alkyne is an excellent building block for developing fluoresceinated bioconjugates by reacting with an azide group.
- Multiple Bioconjugation Applications
- Excellent Quality
Precaución: For Research Use Only
MW:
684.75 g/mol |
CAS Number:
478801-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...