6-FAM Azide ≥95%
Supplier: AAT BIOQUEST INC
133 955
76480-608EA 188.87 USD
76480-608 76480-610
6-FAM Azide ≥95%
6-FAM Azide
6-FAM azide is an excellent building block for developing fluoresceinated bioconjugates by reacting with an alkyne group.
  • Multiple Bioconjugation Applications
  • Excellent Quality
Precaución: For Research Use Only
CAS Number: 1386385-76-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff0fcbf5793ea270032374cd9fd8dbb815a5dddb&vid=b6b554cb05880a9ab29be658b5263a5bfd178dd0&prid=web&clid=SAPDAM