5-FAM-Alkyne ≥95%

Supplier: AAT BIOQUEST INC

Synonyms: 3',6'-Dihydroxy-3-oxo-N-(prop-2-yn-1-yl)-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxamide

132 951
76480-604EA 188.87 USD
76480-604 76480-606
5-FAM-Alkyne ≥95%
5-FAM-Alkyne

5-FAM alkyne is an excellent building block for developing fluoresceinated bioconjugates by reacting with an azide group.


  • Multiple Bioconjugation Applications
  • Excellent Quality


Precaución: For Research Use Only

Formula: C₂₄H₁₅NO₆
MDL Number: MFCD28505571
CAS Number: 510758-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f45ad798e3968276c1ffe8b1f717b48066f8090c&vid=87b09c16fe8c74526fec2ae1b5337a856af13947&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR