5-FAM azide ≥95%
Supplier: AAT BIOQUEST INC
131 950
76480-600EA 188.87 USD
76480-600 76480-602
5-FAM azide ≥95%
5-FAM azide
5-FAM azide is an excellent building block for developing fluoresceinated bioconjugates by reacting with an alkyne group.
  • Multiple Bioconjugation Applications
  • Excellent Quality
Precaución: For Research Use Only
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7aac07f5ffe8cc3a0f4e03febde23801415192a&vid=6457ed13074836d995c602234654e221eb07b914&prid=web&clid=SAPDAM