Rat PPARd Luciferase Reporter Assay Kit
Supplier: INDIGO BIOSCIENCES INC MS
R00121-32 R00121
103881-698KT 1255.38 USD
103881-698 103881-700
Rat PPARd Luciferase Reporter Assay Kit
Assays Cellular Assays
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO’s Rat Peroxisome Proliferator-Activated Receptor delta reporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert against rPPARd (NR1C2).
  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, Luciferase Detection Reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)
This is an all-inclusive cell-based luciferase reporter assay kit targeting the Rat Peroxisome Proliferator-Activated Receptor Delta. INDIGO's rPPAR delta reporter assay utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of the rPPAR delta. In addition to rPPAR delta Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, a reference agonist, Luciferase Detection Reagent, and a cell culture-ready assay plate. The principal application of this assay is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against rPPAR delta. This kit provides researchers with: Clear, reproducible results, exceptional cell viability post-thaw, consistent results lot to lot.
Información para pedidos: Kits are all-inclusive assay systems featuring live cells.
Información de suministro: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C.
Precaución: Do not store in liquid nitrogen. Note reporter cells cannot be refrozen or maintained in extended culture.