Human RARa Luciferase Reporter Assay Kit
Supplier: INDIGO BIOSCIENCES INC MS
IB02202 IB02001-32 IB02001
MSPP-IB02202EA 2929.23 USD
MSPP-IB02202 103881-590 103881-592
Human RARa Luciferase Reporter Assay Kit
Assays Cellular Assays
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO’s Human Retinoic Acid Receptor alphareporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert against RARa (NR1B1).
  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, Luciferase Detection Reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)
This is an all-inclusive cell-based luciferase reporter assay kit targeting the Human Retinoic Acid Receptor alpha (RARa). INDIGO's RAR Alpha reporter assay utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of the RAR Alpha. In addition to RAR Alpha Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, a reference agonist, Luciferase Detection Reagent, and a cell culture-ready assay plate. The principal application of this assay is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against human RAR Alpha. This kit provides researchers with: Clear, reproducible results, exceptional cell viability post-thaw, consistent results lot to lot.
Información para pedidos: Kits are all-inclusive assay systems featuring live cells.
Información de suministro: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at –80 °C. Do not store in liquid nitrogen.
Precaución: Do not store in liquid nitrogen. Note reporter cells cannot be refrozen or maintained in extended culture.