Live Cell Multiplex Cytotoxicity Assay Kit
Supplier: INDIGO BIOSCIENCES INC MS
LCM-01 LCM-05 LCM-10
103881-816KT 227.69 USD
103881-816 103881-818 103881-820
Live Cell Multiplex Cytotoxicity Assay Kit
Assays Cellular Assays
Live Cell Multiplex (LCM) cytotoxicity assay kit.
  • Easy to Understand Viabiltiy Data
  • Run in Conjunction with INDIGO Assays
  • Data in Under 24 Hours
The Live Cell Multiplex (LCM) Assay provides an efficient fluorescence-based method of quantifying the relative number of live cells resident in treated wells of an assay plate. While the LCM Assay may be performed as a stand-alone assay, it has been specifically optimized to be run in multiplex with any of INDIGO’s 96-well, 2×48-well, or 3×32-well Reporter Assay System products.
The LCM assay allows users to validate their primary assay data by determining if their test compound treatments exert mitogenic, cytostatic or cytotoxic activities on reporter cells. The occurrence of such adverse non-specific effects will always undermine the accurate assessment of a test compound’s potency and/or efficacy as a modulator of nuclear receptor function.
Información para pedidos: Kits are all-inclusive assay systems featuring live cells.
Información de suministro: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C. Do not store in liquid nitrogen.