Disposable Dissecting Boards, Mortech®
Supplier: Mortech Manufacturing
BC007 BC008 BC009
76498-294EA 240.01 USD
76498-294 76498-296 76498-298
Disposable Dissecting Boards, Mortech®
Dissecting Boards
Disposable dissection boards are ideal for a wide variety of pathology applications including photography, specimen isolation, specimen pinning, and satellite operations.
  • Multiple sizes and pack quantities available
Cutting surface offers standard and metric scales and sizing grids.