Necrostatin-1 ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-((1H-Indol-3-yl)methyl)-3-methyl-2-thioxoimidazolidin-4-one, Nec-1

A181851-100MG A181851-10MG A181851-1MG A181851-50MG A181851-5MG A181851-250MG A181851-1G
76720-902EA 187.78 USD
76720-902 76720-904 76720-906 76720-908 76720-910 77600-468 77667-508
Necrostatin-1 ≥98%
Necrostatin-1

For Research Use Only

Formula: C₁₃H₁₃N₃OS
MW: 259.327 g/mol
MDL Number: MFCD00056916
CAS Number: 4311-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR