SARS-CoV-2 Spike S1-coupled Magnetic Beads, Acro Biosystems
Supplier: ACROBIOSYSTEMS INC MS
MBSK00110MG MBSK0012MG
103871-546EA 2773.08 USD
103871-546 103871-548
SARS-CoV-2 Spike S1-coupled Magnetic Beads, Acro Biosystems
Beads Magnetic Beads
This product is intended for immunocapture, biopanning and flow cytometry. This product is produced non sterile.
The SARS-CoV-2 Spike S1-coupled Magnetic Beads is produced by coupling biotinylated SARS-CoV-2 Spike S1 protein to streptavidin-conjugated magnetic beads. The pre-coupled beads are ready to use for capturing anti- SARS-CoV-2 antibody or ACE2 protein from your sample with high specificity. The uniform size and large surface-to-volume ratio of the beads ensure highly efficent immunocapture in a simple, fast and convenient workflow with high reproducibilty of data.
Application Method:
a) Reconstitute the Beads following the COA, wash the Beads and suspended to a certain concentration by adding assay Buffer.
b) Add your antibody or ACE-2 protein to the beads, incubation and wash the beads.
Certificaciones: ISO 9001 and ISO 13485.