Human BCMA-coupled Magnetic Beads, Acro Biosystems
Supplier: ACROBIOSYSTEMS INC MS
MBSK00410MG MBSK0042MG
103871-342EA 2773.08 USD
103871-342 103871-344
Human BCMA-coupled Magnetic Beads, Acro Biosystems
Beads Magnetic Beads
This product is intended for immunocapture, biopanning and flow cytometry. This product is produced non sterile.
The Human BCMA-coupled Magnetic Beads is produced by coupling biotinylated Human BCMA protein to streptavidin-conjugated magnetic beads. The pre-coupled beads are ready to use for capturing anti-BCMA antibody from your sample with high specificity. The uniform size and large surface-to-volume ratio of the beads ensure highly efficent immunocapture in a simple, fast and convenient workflow with high reproducibilty of data.
Application Method:
a) Reconstitute the Beads following the COA. Wash and re-suspended the beads to a certain concentration by adding your dilution buffer.
b) Add the prepared beads to your samples.
c) Beads can be separated from your samples afterwards using a magnetic plate.
Certificaciones: ISO 9001 and ISO 13485.