Human EpCAM-coupled Magnetic Beads, Acro Biosystems
Supplier: ACROBIOSYSTEMS INC MS
MBEK01110MG MBEK0112MG
103871-272EA 2315.38 USD
103871-272 103871-274
Human EpCAM-coupled Magnetic Beads, Acro Biosystems
Beads Magnetic Beads
This product is intended for immunocapture, biopanning and flow cytometry. This product is produced non sterile.
The Human EpCAM-coupled Magnetic Beads is produced by coupling biotinylated human EpCAM protein to streptavidin-conjugated magnetic beads. The pre-coupled beads are ready to use for capturing anti-EpCAM antibody from your sample with high specificity. The uniform size and large surface-to-volume ratio of the beads ensure highly efficent immunocapture in a simple, fast and convenient workflow with high reproducibility of data.
Application Method:
a) Reconstitute the BeadsMore Product Information