Human EGFR-coupled Magnetic Beads, Acro Biosystems
Supplier: ACROBIOSYSTEMS INC MS
MBEK01210MG MBEK0122MG
103871-268EA 2773.08 USD
103871-268 103871-270
Human EGFR-coupled Magnetic Beads, Acro Biosystems
Beads Magnetic Beads
This product is intended for immunocapture, biopanning and flow cytometry. This product is produced non sterile.
The Human EGFR-coupled Magnetic Beads is produced by coupling biotinylated human EGFR protein to streptavidin-conjugated magnetic beads. The pre-coupled beads are ready to use for capturing anti-EGFR antibody from your sample with high specificity. The uniform size and large surface-to-volume ratio of the beads ensure highly efficent immunocapture in a simple, fast and convenient workflow with high reproducibility of data.
Application Method:
a) Reconstitute the BeadsMore Product Information