PermaSleeve® Polyolefin Wire Marking Sleeves, Brady
Supplier: Brady Worldwide
PS-C-125-WT PS-C-187-WT PS-C-250-WT PS-C-250-WT-J PS-C-375-WT-J PS-C-187-YL PS-C-094-WT PS-C-500-WT PS-C-500-WT-J PS-C-750-WT
76460-984RL 119.22 USD
76460-984 76460-986 76460-988 76460-990 76460-992 76460-994 76460-996 76460-998 76461-000 76461-002
PermaSleeve® Polyolefin Wire Marking Sleeves, Brady
Labels
PermaSleeve® Heat Shrink Tubing is engineered for thermal transfer or dot matrix printing and is ideal for use in wire identification.
  • Flame-retardant
  • Heat-shrinkable
  • Wide variety of wire diameter ranges
This part is required to be externally fed into most thermal transfer printer models and will not physically mount inside the printer. When used with BradyPrinter i5100 and BRADY IP® Series Label Printer this part will also operate in full manual mode and is not enabled with Brady PrintSmart chip technology. For use with other printers, consult printer's specification for minimum media width and material type compatability, and test for intended use. For use with other printers, this part may also be required to be externally fed.
Sleeves are designed for use with BBP®12 Label Printer, BBP®72 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.
Información para pedidos: A ribbon is required to print this material. Use Part Number IP-R6606 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use R6606 for a different printer.