(2,4-Dichlorophenoxy)acetic acid sodium salt monohydrate ≥98% (by GC), BioReagent suitable for plant cell culture, Sigma-Aldrich®

Danger

Synonyms: 2,4-D sodium salt monohydrate

M-Clarity™ quality level = MQ200

D6679-25G D6679-5G
220122-795EA 651.88 USD
220122-795 220122-796
(2,4-Dichlorophenoxy)acetic acid sodium salt monohydrate ≥98% (by GC), BioReagent suitable for plant cell culture, Sigma-Aldrich®
(2,4-Dichlorophenoxy)acetic acid sodium salt monohydrate
Formula: C₈H₅Cl₂NaO₃·H₂O
MW: 261.03 g/mol
MDL Number: MFCD01311802
CAS Number: 7084-86-8
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance (Color) White to Off-White
Appearance (Form) Powder
Solubility (Color) Colorless
Solubility (Turbidity, 50 mg/ml, H₂O) Clear
Water (by Karl Fischer) ≤10 %
Carbon (anhydrous) 37.5 - 41.3 %
Sodium (Na) anhydrous 7.1 - 11.8 %
Purity (GC) ≥98 %
Plant Cell Culture Test Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR