NEBNext® Globin and rRNA Depletion Kit (Human/Mouse/Rat), New England Biolabs
Supplier: New England Biolabs (NEB)
E7750L E7750S E7750X
76377-488EA 1285 USD
76377-488 76377-490 76377-492
NEBNext® Globin and rRNA Depletion Kit (Human/Mouse/Rat), New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext Globin and rRNA Depletion Kit (Human/Mouse/Rat) employs the NEBNext RNase H-based RNA depletion workflow to deplete adult, embryonic and fetal globin mRNA (HBA1/2, HBB, HBD, HBM, HBG1/2, HBE1, HBQ1 and HBZ), cytoplasmic rRNA (5S, 5.8S, 18S, 28S, ITS and ETS) and mitochondrial rRNA (12S and 16S).
  • Efficient, specific depletion of rRNA and globin mRNA (adult, fetal and embryonic)
  • Suitable for low-quality or high-quality RNA
  • Compatible with a broad range of input amounts: 10 ng - 1 ug
  • Optional integration with poly(A) mRNA isolation workflows for removal of globin RNAs, rRNAs, and noncoding RNAs
  • Fast workflow: 2 hours, with less than 10 minutes hands-on time
The kit is effective with human, mouse and rat total RNA preparations, both intact and degraded. The resulting depleted RNA is suitable for RNA-seq, random-primed cDNA synthesis, or other downstream RNA analysis. This kit can also be used following poly(A) mRNA enrichment (e.g., using the NEBNext poly(A) mRNA Magnetic Isolation Module NEB #E7490), so that the final depleted RNA contains only mRNA of interest and no non-coding RNA.