Argatroban monohydrate ≥98% (by HPLC), Sigma-Aldrich®

Synonyms: MQPA, (2R,4R)-1-[(2S)-5-[(Aminoiminomethyl)amino]-1-oxo-2-[[(1,2,3,4-tetrahydro-3-methyl-8-quinolinyl)sulfonyl]amino]pentyl]-4-methyl-2-piperidinecarboxylic acid, Argipidine, MQPA, (2R,4R)-1-[(2S)-5-[(Aminoiminomethyl)amino]-1-oxo-2-[[(1,2,3,4-tetrahydro-3-methyl-8-quinolinyl)sulphonyl]amino]pentyl]-4-methyl-2-piperidinecarboxylic acid, (2R,4R)-1-((2S)-5-Guanidino-2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulphonamido)pentanoyl)-4-methylpiperidine-2-carboxylic acid hydrate, Argipidine, Argatroban Monohydrate

M-Clarity™ quality level = MQ100

A0487-5MG A0487-25MG
102325-448EA 109.75 USD
102325-448 102341-834
Argatroban monohydrate ≥98% (by HPLC), Sigma-Aldrich®
Argatroban monohydrate

Argatroban anticoagulant is synthesized from L-arginine and photodegrades under light exposure.


Application
Argatroban monohydrate has been used as a thrombin-selective inhibitor in the thrombin assay of platelet poor plasma samples. It may be used as a standard in gradient reversed-phase liquid chromatography coupled with high-resolution multistage mass spectrometry (LC/HR-MSn) for photoproducts fragmentation analysis and as a thrombin inhibitor competitive assay with fullerene-based C60 nanoparticle (NPs).

Formula: C₂₃H₃₆N₆O₅S·H₂O
MW: 526.65 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00895735
CAS Number: 141396-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR