4,5,6-Triaminopyrimidine sulfate Hydrate ≥98%, Sigma-Aldrich®

Warning

Synonyms: Pyrimidine-4,5,6-triamine sulphate

M-Clarity™ quality level = MQ100

307181-1G 307181-5G
220053-601EA 123.85 USD
220053-601 220053-602
4,5,6-Triaminopyrimidine sulfate Hydrate ≥98%, Sigma-Aldrich®
4,5,6-Triaminopyrimidine sulfate Hydrate

4,5,6-Triaminopyrimidine sulfate was converted to 4,5,6-triaminopyrimidine sulfite by formylation with C13 labeled formic acid that was used in the preparation of adenine labeled with C13.

Formula: C₄H₇N₅·H₂SO₄
MW: 223.21 g/mol
MDL Number: MFCD00012789
CAS Number: 6640-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR